Learn More
Chem-Impex International, Inc.Supplier Diversity Partner L-Valine tert-butyl ester hydrochloride, 13518-40-6, MFCD00034863, 25g DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0316825G
Molecular Formula C9H19NO2·HCl, Purity Limit ≥ 99% (HPLC), Molecular Weight 209.7, Melting Point 138-146 ºC, Synonyms: L-Val-OtBu·HCl
Catalog No. 50-497-357