Learn More
AdipoGenSupplier Diversity Partner H1152(10MG) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner SYN1221M010
H-1152, AdipoGen Life Sciences. Chemical. CAS: 871543-07-6. Formula: C16H21N3O2S . 2HCl. MW: 319.4 . 72.9. H-1152 is a potent, specific, ATP-competitive and cell permeable ROCK inhibitor, with an IC50 of 12nM for ROCKII. H-1152 poorly inhibits PKA, PKC and myosin light chain kinase. Rho kinase (ROCK) activated by GTP-linked Rho phosphorylates targets that are involved in cytoskeletal remodelling, smooth muscle contraction and neuronal development.
Catalog No. 50-114-7537