Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ H13N8 (A/black-headed gull/Netherlands/1/00, AAV91212, 17 a.a. - 530 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Supplier: Abnova™ P4886
Specifications
AAV91212 | |
SDS-PAGE | |
H13N8 (A/black-headed gull/Netherlands/1/00) Recombinant Protein | |
50 ug | |
RUO | |
Human | |
His | |
Liquid |
1 mg/mL | |
Liquid | |
Mammalian cell (293) expression system | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
H13N8 | |
Recombinant | |
Mammalian cell (293) expression system | |
>95% by SDS-PAGE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction