Learn More
Abnova™ H16N3 (A/black-headed gull/Sweden/5/99, AAV91217, 16 a.a. - 530 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P4888
Catalog No. 89-946-948
Add to cart
Specifications
Specifications
AAV91217
1 mg/mL
Liquid
H16N3 (A/black-headed gull/Sweden/5/99) Recombinant Protein
>95% by SDS-PAGE
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
H16N3
Yes
Mammalian cell (293) expression system
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
50 ug
RUO
Human
His