Learn More
Abnova™ H1N1 (A/South Carolina/1/18, AAD17229, 19 a.a. - 529 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P4896
Catalog No. 89-946-966
Add to cart
Specifications
Specifications
AAD17229
1 ug/uL
Liquid
H1N1 (A/South Carolina/1/18) Recombinant Protein
>95% by SDS-PAGE
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
H1N1
Yes
Mammalian cell (293) expression system
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
50 ug
RUO
Human
His