Learn More
Abnova™ H2-ALPHA 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00113457T01
Catalog No. 89-946-978
Add to cart
Specifications
Specifications
Western Blot
293T
NM_080386
pCMV-H2-ALPHA full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
H2-ALPHA 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 50kDa
Transient overexpression cell lysate was tested with anti-H2-ALPHA antibody (Mfr. No. H00113457-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)