Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H2AFB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546161
Description
Peptide sequence: SSGAGGRGRT CSRTVRAELS FSVSQVERSL REGHYAQRLS RTAPVYLAAV Sequence homology: Human: 100%.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene encodes a member of the histone H2A family. This gene is part of a region that is repeated three times on chromosome X, once in intron 22 of the F8 gene and twice closer to the Xq telomere. This record represents the most centromeric copy which is in intron 22 of the F8 gene.
Specifications
H2AFB1 | |
Polyclonal | |
Unconjugated | |
H2AFB1 | |
H2A Barr body-deficient; H2A histone family member B1; H2A histone family, member B1; H2A.B; H2A.Bbd; H2AFB1; Histone H2A-Bbd type 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
474382 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P0C5Y9 | |
H2AFB1 | |
Synthetic peptide directed towards the N-terminal of human H2AFB1 (aa 10-59). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction