Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2AFX Mouse anti-Human, Clone: 980CT1.5.5, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human H2AFX.

Manufacturer:  Abnova Corporation MAB12334

Catalog No. 89-934-224


Add to cart

Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif. [provided by RefSeq]

Specifications

Specifications

H2AFX
980CT1.5.5
Mouse monoclonal antibody raised against synthetic peptide of human H2AFX.
In PBS (0.09% sodium azide)
H2A.X/H2A/X/H2AX
Mouse
IgG1
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
H2AFX
A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-30 at N-terminus region of human H2AFX.
Protein G purification
RUO
Primary
3014
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit