Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2AFY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$496.07

Specifications

Antigen H2AFY2
Concentration 0.32 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-1337
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
170301AP150UL
150 μL
Each of 1 for $496.07
Add to Cart
 
Description

Description

Variant histone H2A which replaces conventional H2A in a subset of nucleosomes where it represses transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in stable X chromosome inactivation.
Specifications

Specifications

H2AFY2
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q8CCK0, Q9P0M6
361844, 404634, 55506
H2AFY2 Fusion Protein Ag10487
Primary
-20°C
H2AFY2
0.32 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Core histone macro H2A.2, H2A histone family, member Y2, H2AFY2, Histone macroH2A2, macroH2A2, mH2A2
H2afy2
IgG
Antigen Affinity Chromatography
Antibody
SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit