Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2BFWT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548787

Catalog No. PA548787

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Atypical histone H2B. Nucleosomes containing it are structurally and dynamically indistinguishable from those containing conventional H2B. However, unlike conventional H2B, does not recruit chromosome condensation factors and does not participate in the assembly of mitotic chromosomes. May be important for telomere function.
Specifications

Specifications

H2BFWT
Polyclonal
PBS with 0.09% sodium azide
Q7Z2G1
Histone H2B type W-T, H2B histone family member W testis-specific, H2BFWT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
158983
Western Blot
Unconjugated
H2BFWT
Liquid
H2BFWT
KLH conjugated synthetic peptide between 62-88 amino acids from the Central region of human H2BFWT.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit