Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H2L 5765834, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Antagonist of the lysophosphatidic acid receptors LPA1, LPA5 and LPA3 (IC50 values are 94, 463 and 752 nM respectively). Exhibits no effect at LPA2 or LPA4 receptors.
Specifications
Quantity | 10 mg |
Formula Weight | 404.33 |
Percent Purity | >99% |
Chemical Name or Material | H2L 5765834 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction