Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H3.3 G34W Rabbit anti-Human, Clone: 4H32L4, Invitrogen™ Non-distribution product as customer accommodation.

Rabbit Recombinant Monoclonal Antibody

$149.00 - $425.00

Specifications

Antigen H3.3 G34W
Clone 4H32L4
Concentration 0.5 mg/mL
Applications ChIP assay, Immunocytochemistry, Immunofluorescence, SNAP-ChIP, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-383-6 Non-distribution product as customer accommodation.
View Documents
N/A 100 μg
Each for $425.00
Add to cart
 
70383620UG Non-distribution product as customer accommodation.
View Documents
N/A 20 μg
Each for $149.00
Add to cart
 
Description

Description

This antibody is predicted to react with Mouse, Rat, Bovine.

H3.3 G34W Recombinant Monoclonal antibody specifically detects H3.3 G34W in Human samples. It is validated for ChIP assay, Immunocytochemistry, Immunofluorescence, SNAP-ChIP, Western Blot
Specifications

Specifications

H3.3 G34W
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.4
H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3; H3.3 G34W; H3.3A; H3.3B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3B; H3FT; H3t; Histone 3.3; Histone cluster 3; Histone H3; Histone H3.3; Histone H3.3 G34W; Histone H3.3G34W; PP781
Peptide corresponding to human H3.3G34W [aa30-aa39]
Monoclonal
Protein A
Antibody
4H32L4
ChIP assay, Immunocytochemistry, Immunofluorescence, SNAP-ChIP, Western Blot
Unconjugated
Rabbit
Human
P84243
3020, 3021
IgG
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
H3-3B
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit