Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H3K27me2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen 491013

Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Specifications
H3K27me2 | |
Polyclonal | |
Unconjugated | |
H3C1 | |
dimethyl H3, di-methyl H3, Dimethyl Histone H3, di-methyl histone H3, Di-Methyl-H3 K27, Di-Methyl-H3 Lys27, Di-Methyl-Histone H3 K27, Di-Methyl-Histone H3 Lys27, Di-Tri-Methyl-Histone H3 K27, Di-Tri-Methyl-Histone H3 Lys27, H3, H3 histone, H3 histone family, member A, H3 histone family, member I, H3 histone family, member K, H3 histone family, member L, H3 histone family, member M, H3 histone, family 2, H3 histone, family 3A, H3 histone, family 3B, H3 histone, family 3B (H3.3B), H3 histone, family 3B.1, H3 histone, family 3C, H3 K27me2, H3.1-221, H3.1-291, H3.1-I, H3.2, H3.2-221, H3.2-614, H3.2-615, H3.2-616, H3.3A, H3.3B, H3.5, H3/A, H3/b, H3/d, H3/f, H3/i, H3/j, H3/k, H3/l, H3/M, H3/n, H3/o, H3-143, H3-291, H3-3A, H3-3B, h3-5, H3-53, H3-614, H3a, H3b, H3-B, H3C1, H3c10, H3c11, H3C12, H3C2, H3C3, H3C4, H3C6, H3C7, H3C8, H3f, H3-F, H3F1K, H3F2, H3F3, h3f3a, H3F3B, h3f3b.1, h3f3c, h3f3d, H3FA, H3FB, H3FC HIST1H3C, H3FD, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, H3FM, H3FN, H3g, H3h, H3i, H3k27, H3K27me2/3, H3K27me2me3, H3Lys27me2, H3Lys27me2/3, H3Lys27me2me3, Hist1, HIST1H3A, HIST1H3B, Hist1h3c, HIST1H3D, Hist1h3e, HIST1H3F, Hist1h3g, HIST1H3H, HIST1H3I, HIST1H3J, HIST2H3A, Hist2h3b, HIST2H3C, Hist2h3c1, Hist2h3c2, Hist2h3c2-ps, Hist2h3ca1, Hist2h3ca2, HIST2H3D, histone 1, H3a, histone 1, H3b, histone 1, H3f, histone 1, H3h, histone 2, H3a, histone 2, H3c, histone 2, H3c2, histone 2, H3ca2, Histone 3, histone cluster 1, H3a, histone cluster 1, H3b, histone cluster 1, H3f, histone cluster 1, H3h, histone cluster 2, H3a, histone cluster 2, H3c, histone cluster 2, H3c2, histone cluster 2, H3c2, pseudogene, histone gene complex 1, histone H3, histone H3.1, Histone H3.2, histone H3.3, Histone H3.3C, histone H3.5, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, histone H3/l, Histone H3/m, histone H3/o, Histone H3K27, Histone H3K27me2, histone variant H3.5, hypothetical protein LOC550262, M32461, methyl Histone 3, methyl Histone H3, PP781, wu:fa25h06, wu:fa96g06, wu:fb07a08, wu:fb36f01, zgc:110292, zgc:174300, zgc:56193, zgc:56418, zgc:64222, zgc:86731 | |
Rabbit | |
Affinity chromatography | |
RUO | |
126961, 333932, 550262, 8350, 8356, 8357, 8358, 8968 | |
-20°, C, Avoid Freeze/Thaw Cycles | |
IgG |
Dot Blot, ELISA, Immunocytochemistry, Western Blot | |
2.63 mg/mL | |
PBS with 0.05% ProClin 300, 0.05% sodium azide | |
P68431, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, h3f3b.1 | |
Raised against the region of the histone H3 containing the dimethylated lysine 27 (H3K27me2), using a KLH-conjugated synthetic peptide. | |
50 μg | |
Primary | |
Human, Zebrafish | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction