Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H3K4me3Q5dopaminyl Rabbit anti-Rat, Human, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$145.80 - $373.00

Specifications

Antigen H3K4me3Q5dopaminyl
Concentration 0.03 mg/mL
Applications Dot Blot, Inhibition Assays, Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABE2590100
View Documents
MilliporeSigma
ABE2590100UL
100 μL
Each for $373.00
Add to cart
 
ABE259025UL
View Documents
MilliporeSigma
ABE259025UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

H3K4me3Q5dopaminyl Polyclonal antibody specifically detects H3K4me3Q5dopaminyl in Rat, Human samples. It is validated for Dot Blot, Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

H3K4me3Q5dopaminyl
Dot Blot, Inhibition Assays, Western Blot
Purified
RUO
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
0.03 mg/mL
Unconjugated
Rabbit
Rat, Human
KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3 with trimethylated lysine 4 and dopaminylated on glutamine 5.
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit