Learn More
H3K79me2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific 720079
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. Since it is conserved across species, the antibody may react with many other species.
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post tranlationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Specifications
Di-Methyl-Histone H3 (Lys79) | |
Polyclonal | |
Unconjugated | |
HIST1H3A | |
H3K79me2, Histone H3, Histone 1, H3a, Histone H3.1, histone H3/a, HIST1H3A | |
Rabbit | |
Affinity chromatography | |
RUO | |
8350 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP assay, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
P68431 | |
HIST1H3A | |
Methylated peptide (Lys79) corresponding to Human H3 (aa 75-83) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.