Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H4K20me3 Recombinant Polyclonal Antibody (37HCLC), ChIP-Verified, Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Supplier: LIFE TECHNOLOGIES 71285520UG
Description
This antibody is predicted to react with Mouse.
H4K20me3 Recombinant Polyclonal antibody specifically detects H4K20me3 in Human samples. It is validated for ChIP Assay.Specifications
H4K20me3 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P62805 | |
H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9 | |
Peptide corresponding to human H4K20me3 [aa17-aa25] | |
20 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay | |
37HCLC | |
Unconjugated | |
H4C1 | |
CELE_T10C6.11; F07B7.11; F07B7.4; F45F2.12; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K20; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4c11; H4C12; H4C13; H4c14; H4C15; H4C2; H4C3; H4C4; H4C5; H4c6; H4C8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K20; H4Lys20me3; H4M; his-20; his-22; his-4; his-52; his-54; his-8; HIST1H4; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; Hist1h4h; Hist1h4i; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone c | |
Rabbit | |
Protein A | |
RUO | |
121504, 8294, 8359, 8363, 8366, 8367, 8370 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction