Learn More
Abnova™ H5 (H5N1) (A/Anhui/1/2005) (ABD28180, 18 a.a. - 530 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5062
Catalog No. 89-946-998
Add to cart
Specifications
Specifications
ABD28180
1 ug/uL
Liquid
H5 (H5N1) (A/Anhui/1/2005) Recombinant Protein
≥98% by SDS-PAGE
50 ug
RUO
Human
His
Mammalian cell (293) expression system
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
H5
Yes