Learn More
Abnova™ H5 (H5N1) (A/Egypt/Chicken/2253-1/2006 (ABG81039, 17 a.a. - 530 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5068
Catalog No. 89-946-994
Add to cart
Specifications
Specifications
ABG81039
1 ug/uL
Liquid
H5 (H5N1) (A/Egypt/Chicken/2253-1/2006 Recombinant Protein
≥95% by SDS-PAGE
50 ug
RUO
Human
His
Mammalian cell (293) expression system
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
H5
Yes