Learn More
Abnova™ H5 (H5N1) (A/Japanese white-eye/Hong Kong/1038/2006) (ABJ96775, 17 a.a. - 530 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5070
Catalog No. 89-946-940
Add to cart
Specifications
Specifications
ABJ96775
1 ug/uL
Liquid
H5 (H5N1) (A/Japanese white-eye/Hong Kong/1038/2006) Recombinant Protein
≥95% by SDS-PAGE
50 ug
RUO
Human
His
Mammalian cell (293) expression system
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
H
Yes