Learn More
Abnova™ H5 (H5N1) (A/Vietnam/1203/2004) (AAW80717, 18 a.a. - 530 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5060
Catalog No. 89-947-000
Add to cart
Specifications
Specifications
Mammalian cell (293) expression system
1 ug/uL
Liquid
H5 (H5N1) (A/Vietnam/1203/2004) Recombinant Protein
≥98% by SDS-PAGE
50 ug
RUO
Human
His
AAW80717
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
H5
Yes