Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H60 Antibody (RM0248-11G35), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212428AF532
Description
H60 Monoclonal specifically detects H60 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
H60 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Rat | |
Protein A or G purified | |
Cancer | |
15101.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
RM0248-11G35 | |
Western Blot, Flow Cytometry | |
H60a | |
Mouse recombinant protein of H60. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction