Learn More
H60 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0248-11G35, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212428AF594
Catalog No. NP212428A94
Add to cart
Description
Description
H60 Monoclonal antibody specifically detects H60 in Mouse samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
H60
RM0248-11G35
Western Blot, Flow Cytometry
H60a
Mouse recombinant protein of H60.
0.1 mL
Primary
15101
Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse