Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA 1100 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
155558-32-0
C14H18ClN3O3S
343.826
XWWFOUVDVJGNNG-UHFFFAOYSA-N
hydroxyfasudil hydrochloride,hydroxyfasudil hydrochloride,ha1100 hydrochloride,ha 1100 hydrochloride,hydroxy fasudil hydrochloride,1-1-hydroxy-5-isoquinolinesulfonyl homopiperazine, hcl,hydroxyfasudil hcl,ha-1100 hydrochloride,hydroxyfasudil hydrochloride,c14h17n3o3s.clh
11371328
5-(1,4-diazepan-1-ylsulfonyl)-2H-isoquinolin-1-one;hydrochloride
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CNC3=O.Cl
This item is not returnable. View return policy
155558-32-0
C14H18ClN3O3S
343.826
XWWFOUVDVJGNNG-UHFFFAOYSA-N
hydroxyfasudil hydrochloride,hydroxyfasudil hydrochloride,ha1100 hydrochloride,ha 1100 hydrochloride,hydroxy fasudil hydrochloride,1-1-hydroxy-5-isoquinolinesulfonyl homopiperazine, hcl,hydroxyfasudil hcl,ha-1100 hydrochloride,hydroxyfasudil hydrochloride,c14h17n3o3s.clh
11371328
5-(1,4-diazepan-1-ylsulfonyl)-2H-isoquinolin-1-one;hydrochloride
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CNC3=O.Cl

Cell-permeable, selective Rho-kinase inhibitor

Cell-permeable active metabolite of Fasudil (HA 1077; Cat. No. 0541). Produces ATP-competitive and reversible inhibition of Rho-kinase and is ∽ 100-fold selective over a range of other protein kinases. Inhibits neutrophil migration and produces potent vasodilatory effects in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 343.83
Percent Purity >99%
Chemical Name or Material HA 1100 hydrochloride

Recommended Storage : Store at -20°C