Learn More
Abnova™ HA (B/Florida/4/2006) (ACA33493, 17 a.a. - 546 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5073
Catalog No. 89-947-018
Add to cart
Specifications
Specifications
Mammalian cell (293) expression system
1 mg/mL
Liquid
HA (B/Florida/4/2006) Recombinant Protein
≥95% by SDS-PAGE
50 ug
RUO
Human
His
ACA33493
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
HA
Yes