Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Recombinant Influenza A Virus Protein, subtype H1N1, (A/California/04/2009), His Tag Non-distribution product as customer accommodation.

HA Recombinant Influenza A Virus Protein, subtype H1N1, (A/California/04/2009), His Tag

Manufacturer:  LIFE TECHNOLOGIES 11055V08H50

 View more versions of this product

Catalog No. 11-055-V08H50


Add to cart

Description

Description

Hemagglutinin, Influenza A virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 529) of influenza A virus, subtype H1N1, (A/California/04/2009), HA (ACP41105.1) fused to a polyhistidine tag at the C-terminus. Subunits HA1 and HA2 remain uncleaved.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein assays and analysis
Lyophilized
HA Recombinant Influenza A Virus Subtype H1N1 (A/California/04/2009) Protein
50μg
For research use only. Not for use in diagnostic procedures
Viral proteins
HA Recombinant Influenza A Virus Subtype H1N1 (A/California/04/2009) Protein
Assay standard
Mammalian
Influenza
Lyophilized
59kDa
≥85% by SDS-PAGE
–20 to –70°C
1 Each
HA
HA
Other proteins
HA Recombinant Influenza A Virus Subtype H1N1 (A/California/04/2009) Protein
His Tag
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.