Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA-Tag Mouse anti-Human, Active Motif™ Non-distribution product as customer accommodation.

Mouse Recombinant Monoclonal Antibody

$98.40 - $465.85

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen HA-Tag
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3792 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91272
10 μg
Each for $98.40
Add to cart
 
50-199-3791 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91271
100 μg
Each for $465.85
Add to cart
 
Description

Description

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
Specifications

Specifications

Monoclonal
Primary
HA-Tag
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit