Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Mouse anti-Nonspecies specific, DyLight 650, Clone: 4810, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243714C

Catalog No. NBP243714C


Add to cart

Description

Description

HA Tag Monoclonal antibody specifically detects HA Tag in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

HA Tag
4810
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG2a
0.1 ml
Primary
Nonspecies specific
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
HA epitope tag, Hemagglutinin
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit