Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HA Tag Polyclonal Antibody, eBioscience™, Invitrogen™

Catalog No. 5013532
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-135-32 10 μg
50-135-33 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-135-32 Supplier Invitrogen™ Supplier No. 14675663
  • $155.00
    $116.25 / Each of 1

    Save $38.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Description: The polyclonal antibody reacts with HA tag. One of the more established epitope tags is the HA tag, which is a nine amino acid peptide sequence (YPYDVPDYA) present in the human influenza virus hemagglutinin protein. The HA tag has been successfully used in a variety of applications, including Western blotting, immunoprecipitation, and immunfluorescence studies. Applications Reported: Purified Anti-HA tag poly has been reported for use in immunoprecipitation, and immunoblotting (WB). Applications Tested: Purified Anti-HA tag poly has been tested by immunoblotting (WB). (1:200 starting dilution). It is recommended that this antibody be titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA Tag
Applications ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with BSA, 0.02% gelatin and 0.1% sodium azide
Gene Alias HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.