Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA tag Rabbit, Clone: 109B2, GenScript™

Catalog No. 502120685
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0685 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0685 Supplier Genscript Corporation Supplier No. A0196340
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. The rabbit immune system generates antibody diversity and optimizes affinity. GenScript utilizes MonoRabTM technology to generate high affinity and specific monoclonal rabbit antibodies. The HA tag is composed of several amino acids containing YPYDVPDYA and does not appear to interfere with the bioactivity or the biodistribution of recombinant proteins. GenScript MonoRabTM HA tag Antibody (109B2), mAb, Rabbit is specific to HA tags placed at the N-terminal, C-terminal, and internal regions of fusion proteins. This antibody can greatly improve the effectiveness of several different kinds of immunoassays, helping researchers identify and detect HA-tagged fusion proteins in bacteria, insect cells, and mammalian cells.

HA tag Monoclonal antibody specifically detects HA tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Western Blot

Specifications

Antigen HA tag
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 109B2
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
Host Species Rabbit
Immunogen a synthetic peptide containing the influenza hemagglutinin epitope (YPYDVPDYA). conjugated to KLH
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.