Learn More
Abnova™ HA1 (A/California/06/09) (H1N1) (ACP41935, 18 a.a. - 344 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5040
Catalog No. 89-947-021
Add to cart
Specifications
Specifications
Mammalian cell (293) expression system
1 ug/uL
Liquid
HA1 (A/California/06/09) (H1N1) Recombinant Protein
≥95% by SDS-PAGE
50 ug
RUO
Human
His
ACP41935
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
HA1
Yes