Learn More
Abnova™ HA1 (H1N1) (PR/8/34) (P03452, 18 a.a. - 345 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P5054
Catalog No. 89-947-029
Add to cart
Specifications
Specifications
Mammalian cell (293) expression system
1 ug/uL
Liquid
HA1 (H1N1) (PR/8/34) Recombinant Protein
≥95% by SDS-PAGE
50 ug
RUO
Human
His
P03452
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
HA1
Yes