Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ HA1 (H2N2) (A/Canada/720/2005) (AAY28987, 13 a.a. - 340 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Supplier: Abnova™ P5044
Specifications
AAY28987 | |
SDS-PAGE | |
HA1 (H2N2) (A/Canada/720/2005) Recombinant Protein | |
SDS-PAGE Result | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
HA1 | |
Recombinant | |
Mammalian cell (293) expression system | |
>95% by SDS-PAGE |
1 ug/uL | |
Liquid | |
Mammalian cell (293) expression system | |
50 ug | |
RUO | |
Human | |
His | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction