Learn More
HA14-1, Tocris Bioscience™
Bcl-2 inhibitor. Induces apoptosis
Manufacturer:  Tocris Bioscience 1541/25
 View more versions of this product
Catalog No. 15-412-5
Add to cart
This item is not returnable. View return policy
Description
Description
Cell-permeable inhibitor of Bcl-2 protein (IC50 ~ 9 µM); acts by binding to the surface pocket. Disrupts Bax/Bcl-2 interaction and induces apoptosis of tumor cells. Also binds to the antiapoptotic Blc-2 proteins Bcl-XL and Bcl-w.
Specifications
Specifications
2-Amino-6-bromo-α-cyano-3-(ethoxycarbonyl)-4H-1-benzopyran-4-acetic acid ethyl ester
C17H17BrN2O5
25mg
65673-63-4
Observed MW: 409.23
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at -20°C