Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HABP1/C1QBP/GC1q R Antibody (001), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290475APC
Description
HABP1/C1QBP/GC1q R Monoclonal antibody specifically detects HABP1/C1QBP/GC1q R in Mouse samples. It is validated for ELISASpecifications
HABP1/C1QBP/GC1q R | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
001 | |
ELISA | |
C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein | |
Rabbit immunized with purified, recombinant Mouse HABP1/C1QBP/GC1q R (Accession#: NP_031599.2, Leu72-Gln279) | |
0.1 mL | |
Stem Cell Markers | |
708 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction