Learn More
Abnova™ HABP2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003026T01
Catalog No. 89-947-046
Add to cart
Specifications
Specifications
Western Blot
293T
NM_004132.2
pCMV-HABP2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
HABP2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 61.71kDa
Transient overexpression cell lysate was tested with anti-HABP2 antibody (Mfr. No. H00003026-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)