Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HABP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HABP4
Dilution Immunohistochemistry -1:50-1:200, Immunohistochemistry-Paraffin -1:50-1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121468
View Documents
Novus Biologicals
NBP311380200UL
200 μg
Each for $320.71
Add to cart
 
NB121469
View Documents
Novus Biologicals
NBP31138050UL
50 μg
Each for $138.37
Add to cart
 
Description

Description

HABP4 Polyclonal antibody specifically detects HABP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

HABP4
Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
PBS, pH7.0
22927
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry -1:50-1:200, Immunohistochemistry-Paraffin -1:50-1:200
Unconjugated
Rabbit
Human
chromodomain helicase DNA binding protein 3 interacting protein, hyaluronan binding protein 4, IHABP-4, IHABP4MGC825, intracellular antigen detected by monoclonal Ki-1, intracellular hyaluronan-binding protein 4, Ki-1/57, Ki-1/57 intracellular antigen, SERBP1L
E. coli-derived Human HABP4 fragment
Polyclonal
Antigen and protein A Affinity-purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit