Learn More
HACE1 Mouse anti-Human, Clone: OTI4F10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810063
Catalog No. 50-166-9867
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
E3 ubiquitin-protein ligase involved in Golgi membrane fusion and regulation of small GTPases. Acts as a regulator of Golgi membrane dynamics during the cell cycle: recruited to Golgi membrane by Rab proteins and regulates postmitotic Golgi membrane fusion. Acts by mediating ubiquitination during mitotic Golgi disassembly, ubiquitination serving as a signal for Golgi reassembly later, after cell division. Specifically interacts with GTP-bound RAC1, mediating ubiquitination and subsequent degradation of active RAC1, thereby playing a role in host defense against pathogens. May also act as a transcription regulator via its interaction with RARB.
Specifications
Specifications
HACE1
Monoclonal
Unconjugated
HACE1
Lyophilized
HACE1
Full length human recombit protein of human HACE1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI4F10
PBS with 8% trehalose and no preservative; pH 7.3
Q8IYU2
SPPRS
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
57531