Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Hach™ AquaChek™ Dip-and-Read Water Quality Test Strips
Fast, easy, reliable onsite environmental testing
Manufacturer:  Hach Company 611213H
 View more versions of this product
Catalog No. 23-292755
Add to cart
Specifications Description & Specifications
Specifications
Fast, easy, reliable onsite environmental testing like analyzing water, aquacultures, food, or other similar samples
0 to 425ppm
1 or <1 min.
6.4 to 8.8
50 Pack
Dip-and-read
SofChek™ Water Test Strip