Hach™ AquaChek™ Dip-and-Read Water Quality Test Strips

Fast, easy, reliable onsite environmental testing

Manufacturer: HACH COMPANY 651453H

 View more versions of this product

Catalog No. 23-292-758

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Fast, easy, reliable onsite environmental testing like analyzing water, aquacultures, food, or other similar samples
    Quantity 50 Pack
    Range 0 to 10ppm
    Test Method Dip-and-read
    Test Time 1 or <1 min.
    Product Type AquaChek™ Water Test Strip
    pH Range 6.4 to 8.8

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit