Learn More
Hach™ AquaChek™ Dip-and-Read Water Quality Test Strips
Fast, easy, reliable onsite environmental testing
Manufacturer:  Hach Company 711195H
 View more versions of this product
Catalog No. 23-292-763
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Provide chloride readings of both aqueous or solid extractions and also provides fast, easy, reliable onsite environmental testing like analyzing water, aquacultures, food, or other similar samples
30ppm
1 or <1 min.
6.4 to 8.8
40 Pack
Dip-and-read
Quantab™ Water Test Strip