Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM28, Mouse anti-Human, Alexa Fluor 488, Clone: 1004266, R&D Systems
Mouse Monoclonal Antibody
Manufacturer: R&D Systems FAB93252G100UG
This item is not returnable.
View return policy
Description
ADAM28 Monoclonal antibody specifically detects ADAM28 in Human Samples. It is validated for Flow Cytometry.Specifications
ADAM28 | |
1004266 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q0UKQ2 | |
a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, Dtgn1, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCL, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L, Protein-L | |
Chinese Hamster Ovary cell line, CHO-derived human ADAM28, Leu72 & Leu191-Ala623,, Accession # Q0UKQ2 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
10863 | |
Human |
Flow Cytometry | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Purified | |
Mouse | |
IgG1 | |
100 ug | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
Monoclonal | |
Detects human ADAM28 in direct ELISAs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title