Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Alfa Aesar™

Manufacturer: Alfa Aesar 1256718

Quantity 50g

 View more versions of this product

Catalog No. AA1256718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12069-85-1
    Physical Form Powder
    Assay Percent Range 99.5% (Metals basis excluding Zr)
    Molecular Formula CHf
    Formula Weight 190.54
    MDL Number MFCD00016126
    Chemical Name or Material Hafnium carbide

    Specifications

    Physical Form Powder
    Formula Weight 190.54
    Quantity 50g
    Color Gray
    DOT Information DOT Class: 4.1, Packing Group: III
    Density 12.2g/cm3
    EINECS Number 235-114-1
    Odor Odorless
    Assay Percent Range 99.5% (Metals basis excluding Zr)
    Molecular Formula CHf
    MDL Number MFCD00016126
    CAS 12069-85-1
    Health Hazard 1 H228: Flammable solid.
    Health Hazard 2 Take precautionary measures against static discharges.
    Chemical Name or Material Hafnium carbide

    Take precautionary measures against static discharges.

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit