Alfa Aesar™ Hafnium dichloride oxide octahydrate, Puratronic™, 99.998% (metals basis excluding Zr), Zr <1%

Manufacturer: Alfa Aesar™ 1070506

Quantity 5g

 View more versions of this product

Catalog No. AA1070506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14456-34-9
    Linear Formula HfCl2O·8H2O
    Formula Weight 409.52 (265.40 Anhydrous)
    MDL Number MFCD00150417
    UN Number UN3260
    Chemical Name or Material Hafnium dichloride oxide octahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 409.52 (265.40 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Pungent
    Physical Form Powder
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00150417
    UN Number UN3260
    Density 2.2g/mL
    RTECS Number MG4800000
    Linear Formula HfCl2O·8H2O
    CAS 14456-34-9
    Color White
    Quantity 5g
    Chemical Name or Material Hafnium dichloride oxide octahydrate
    Assay 99.998%, (metals basis excluding Zr), Zr <1%
    Grade Puratronic™

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit