Hafnium dichloride oxide octahydrate, White powder, >98% (Metals basis excluding Zr), Alfa Aesar™

For Synthesis of Hafnium (IV) Oxide

Manufacturer: Alfa Aesar 1183322

Quantity 100g

 View more versions of this product

Catalog No. AA1183322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14456-34-9
    Physical Form Powder
    Assay Percent Range >98% (Metals basis excluding Zr)
    Molecular Formula Cl2HfO·8H2O
    Formula Weight 409.52; 265.40 (anhydrous)
    MDL Number MFCD00150417
    Synonym Hafnium oxychloride
    Chemical Name or Material Hafnium dichloride oxide octahydrate

    Specifications

    Physical Form Powder
    Formula Weight 409.52; 265.40 (anhydrous)
    Quantity 100g
    Color White
    DOT Information DOT Class: 8, Packing Group: III
    EINECS Number 237-345-3
    Assay Percent Range >98% (Metals basis excluding Zr)
    Molecular Formula Cl2HfO·8H2O
    Odor Pungent
    MDL Number MFCD00150417
    CAS 14456-34-9
    Synonym Hafnium oxychloride
    Health Hazard 1 H314-H318-H303: Causes severe skin burns and eye damage. Causes serious eye damage. May be harmful if swallowed.
    Chemical Name or Material Hafnium dichloride oxide octahydrate

    H314-H318-H303: Causes severe skin burns and eye damage. Causes serious eye damage. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit