Alfa Aesar™ Hafnium dinitrate oxide, 99.9% (metals basis excluding Zr), Zr <0.1%, 10% w/v aq. soln.

Manufacturer: Alfa Aesar™ 44246AB

Quantity 10mL

 View more versions of this product

Catalog No. AA44246AB

  Check Availability
Add to Cart

    Chemical Identifiers

    Linear Formula HfO(NO3)2
    Formula Weight 318.5
    MDL Number MFCD00798524
    UN Number UN3218
    Chemical Name or Material Hafnium dinitrate oxide

    Specifications

    Formula Weight 318.5
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00798524
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .
    Physical Form Liquid
    Linear Formula HfO(NO3)2
    UN Number UN3218
    Quantity 10mL
    Concentration 10% w/v aq. soln.
    Chemical Name or Material Hafnium dinitrate oxide
    Assay 99.9%, (metals basis excluding Zr), Zr <0.1%

    GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit