Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hafnium foil, 0.75mm (0.03 in.) thick, 99.5% (metals basis excluding Zr), Zr nominal 3%

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
7440-58-6
Hf
178.49
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium, elemental,hafnio,celtium,unii-x71938l1do,and compounds,hsdb 552,powder,foil,atom,wire
23986
CHEBI:33343
hafnium
[Hf]
7440-58-6
Hf
178.49
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium, elemental,hafnio,celtium,unii-x71938l1do,and compounds,hsdb 552,powder,foil,atom,wire
23986
CHEBI:33343
hafnium
[Hf]
Quantity 25 x 25mm
Weight ≈6.1g/25x25mm
Odor Odorless
Physical Form Foil
Percent Purity 99.5%
Chemical Name or Material Hafnium foil, 0.75mm (0.03 in.) thick
Assay (metals basis excluding Zr), Zr nominal 3%

EINECSNumber : 231-166-4

TSCA : Yes

Recommended Storage : Ambient temperatures