Alfa Aesar™ Hafnium(IV) fluoride, 99.9% (metals basis excluding Zr), Zr 2-4%

Manufacturer: Alfa Aesar™ 3639214

Quantity 25g

 View more versions of this product

Catalog No. AA3639214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13709-52-9
    Formula Weight 254.48
    MDL Number MFCD00016127
    UN Number UN3290
    Chemical Name or Material Hafnium(IV) fluoride

    Specifications

    Formula Weight 254.48
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Particle Size 3 to 6mm
    Physical Form Sintered Lump
    MDL Number MFCD00016127
    Quantity 25g
    Health Hazard 1 Danger
    TSCA TSCA
    Sensitivity Moisture Sensitive
    CAS 13709-52-9
    UN Number UN3290
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, CORROSIVE, INORGANIC, N.O.S.
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Hafnium(IV) fluoride
    Assay 99.9%, (metals basis excluding Zr), Zr 2 to 4%

    GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit