Alfa Aesar™ Hafnium(IV) oxide, 99.95%, (metals basis excluding Zr), Zr typically <0.5%

Manufacturer: Alfa Aesar™ 3566606

Quantity 5g

 View more versions of this product

Catalog No. AA3566606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12055-23-1
    Formula Weight 210.49
    MDL Number MFCD00003565
    Merck Index 14,4588
    Chemical Name or Material Hafnium(IV) oxide

    Specifications

    Formula Weight 210.49
    Mesh Size −325 Mesh
    Odor Odorless
    Physical Form Powder
    Merck Index 14,4588
    Melting Point 2774°C
    Density 9.68g/mL
    TSCA TSCA
    MDL Number MFCD00003565
    Quantity 5g
    CAS 12055-23-1
    Chemical Name or Material Hafnium(IV) oxide
    Assay 99.95%, (metals basis excluding Zr), Zr typically <0.5%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit