Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium(IV) oxide, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
12055-23-1
HfO2
12055-23-1
HfO2
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula HfO2
Formula Weight 210.49
Physical Form Powder
Percent Purity 85% (Hf typical)
Grade Trace Metal Basis
Density 9.6800g/mL
Packaging Glass bottle
Total Trace Metal Impurities 100ppm max.
Color White
Quantity 5g
Chemical Name or Material Hafnium(IV) oxide
Show More Show Less