Alfa Aesar™ Hafnium(IV) oxide sputtering target, 101.4mm (4.0 in.) dia x 6.35mm (0.25 in.) thick, 99.95% (metals basis excluding Zr)

Manufacturer: Alfa Aesar™ 44875KS

Quantity 1 each

 View more versions of this product

Catalog No. AA44875KS

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula HfO2
    MDL Number MFCD00003565
    Chemical Name or Material Hafnium (IV) oxide sputtering target, 101.4mm (4.0 in.) dia. x 6.35mm (0.25 in.) thick

    Specifications

    Molecular Formula HfO2
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00003565
    Quantity 1 each
    Chemical Name or Material Hafnium (IV) oxide sputtering target, 101.4mm (4.0 in.) dia. x 6.35mm (0.25 in.) thick
    Assay 99.95%, (metals basis excluding Zr)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit